Allylamine hydrochloride 60% in water

BLDPBD01392817-25GEA 7.3 EUR
BLDPBD01392817-25G
Allylamine hydrochloride 60% in water
Allylamine hydrochloride

Storage condition: Sealed in dry, Room Temperature

Kaava: C₃H₈ClN
MW: 93,5553 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00054329
CAS-numero: 10017-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR