3-(Difluoromethoxy)benzonitrile 99+%

BLDPBD01409371-100EA 682 EUR
BLDPBD01409371-100 BLDPBD01409371-1G BLDPBD01409371-5G BLDPBD01409371-25G
3-(Difluoromethoxy)benzonitrile 99+%
3-(Difluoromethoxy)benzonitrile

Storage condition: Sealed in dry, 2-8°C

Kaava: C₈H₅F₂NO
MW: 169,1282 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00236225
CAS-numero: 97582-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR