4-Chloro-o-anisidine 98%

BLDPBD01421516-1GEA 28.1 EUR
BLDPBD01421516-1G BLDPBD01421516-25G BLDPBD01421516-100 BLDPBD01421516-5G
4-Chloro-o-anisidine 98%
4-Chloro-o-anisidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₇H₈ClNO
MW: 157,5975 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06801386
CAS-numero: 93-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR