4-Bromo-N,N-diethylbenzenesulphonamide 98%

BLDPBD0222-5GEA 87.3 EUR
BLDPBD0222-5G BLDPBD0222-1G BLDPBD0222-250MG
4-Bromo-N,N-diethylbenzenesulphonamide 98%
4-Bromo-N,N-diethylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₄BrNO₂S
MW: 292,1900 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00587586
CAS-numero: 90944-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR