Dimetyyli-5-bromi-isoftalaatti 97%

BLDPBD0433-10GEA 28.1 EUR
BLDPBD0433-10G BLDPBD0433-5G BLDPBD0433-25G BLDPBD0433-100G BLDPBD0433-1G BLDPBD0433-500G
Dimetyyli-5-bromi-isoftalaatti 97%
Dimetyyli-5-bromi-isoftalaatti

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₀H₉BrO₄
MW: 273,08 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00078709
CAS-numero: 51760-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR