tert-Butyl-4-(bromomethyl)benzoate 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD0846-5GEA 32.3 EUR
BLDPBD0846-5G BLDPBD0846-250MG BLDPBD0846-10G BLDPBD0846-1G BLDPBD0846-25G
tert-Butyl-4-(bromomethyl)benzoate 95%
tert-Butyl-4-(bromomethyl)benzoate

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₂H₁₅BrO₂
MW: 271,15 g/mol
Storage Temperature: Jääkaappi
CAS-numero: 108052-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR