4'-Butoxyacetophenone 98%

BLDPBD0876-25GEA 203 EUR
BLDPBD0876-25G BLDPBD0876-100G BLDPBD0876-1G BLDPBD0876-5G BLDPBD0876-0.025G
4'-Butoxyacetophenone 98%
4'-Butoxyacetophenone

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₂H₁₆O₂
MW: 192,26 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00027200
CAS-numero: 5736-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR