2,4-Dimethyl-5-thiazolecarboxylic acid 98%
|
|
BLDPBD10000-25GEA
172
EUR
BLDPBD10000-25G
BLDPBD10000-10G
BLDPBD10000-5G
BLDPBD10000-1G
2,4-Dimethyl-5-thiazolecarboxylic acid 98%
2,4-Dimethyl-5-thiazolecarboxylic acid
Storage condition: Sealed in dry, 2-8°C
Kaava:
C₆H₇NO₂S MW: 157,1900 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD00052943 CAS-numero: 53137-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...