5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone 95%

BLDPBD100665-5GEA 14.6 EUR
BLDPBD100665-5G BLDPBD100665-500G BLDPBD100665-25G BLDPBD100665-100G
5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone 95%
5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₇H₂₀N₄O₂
MW: 312,37 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03093929
CAS-numero: 139756-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR