5-(2-Nitrophenyl)oxazole 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD100723-5GEA 281 EUR
BLDPBD100723-5G BLDPBD100723-250MG BLDPBD100723-1G
5-(2-Nitrophenyl)oxazole 97%
5-(2-Nitrophenyl)oxazole

Storage condition: Sealed in dry, 2-8°C

Kaava: C₉H₆N₂O₃
MW: 190,1555 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00085146
CAS-numero: 89808-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR