5-(Phenylethynyl)nicotinic acid 95%
|
|
BLDPBD100733-5GEA
698
EUR
BLDPBD100733-5G
BLDPBD100733-100MG
BLDPBD100733-1G
BLDPBD100733-250MG
5-(Phenylethynyl)nicotinic acid 95%
5-(Phenylethynyl)nicotinic acid
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₁₄H₉NO₂ MW: 223,227 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00174055 CAS-numero: 175203-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...