5-Bromo-2-iodobenzonitrile 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD102759-5GEA 34.3 EUR
BLDPBD102759-5G BLDPBD102759-25G BLDPBD102759-10G BLDPBD102759-1G
5-Bromo-2-iodobenzonitrile 95%
5-Bromo-2-iodobenzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₇H₃BrIN
MW: 307,92 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00672950
CAS-numero: 121554-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR