2,3-Difluoro-6-nitroanisole 98%

BLDPBD10361-25GEA 38.5 EUR
BLDPBD10361-25G BLDPBD10361-100G BLDPBD10361-1G BLDPBD10361-5G BLDPBD10361-10G
2,3-Difluoro-6-nitroanisole 98%
2,3-Difluoro-6-nitroanisole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₅F₂NO₃
MW: 189,1163 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03840468
CAS-numero: 66684-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR