5-Amino-1,3-dihydro-2H-benzimidazol-2-one 98%

BLDPBD10390-100GEA 16.7 EUR
BLDPBD10390-100G BLDPBD10390-25G
5-Amino-1,3-dihydro-2H-benzimidazol-2-one 98%
5-Amino-1,3-dihydro-2H-benzimidazol-2-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₇H₇N₃O
MW: 149,1500 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00053555
CAS-numero: 95-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR