4-Chloro-5-methylbenzene-1,2-diamine 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD10587-1GEA 31.2 EUR
BLDPBD10587-1G BLDPBD10587-5G
4-Chloro-5-methylbenzene-1,2-diamine 98%
4-Chloro-5-methylbenzene-1,2-diamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₇H₉ClN₂
MW: 156,613 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00221471
CAS-numero: 63155-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR