2-Chloro-4-methylpyrimidine-5-carboxylic acid 97%

BLDPBD107360-5GEA 1380 EUR
BLDPBD107360-5G BLDPBD107360-100MG BLDPBD107360-1G BLDPBD107360-250MG
2-Chloro-4-methylpyrimidine-5-carboxylic acid 97%
2-Chloro-4-methylpyrimidine-5-carboxylic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₆H₅ClN₂O₂
MW: 172,569 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09863167
CAS-numero: 188781-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR