4'-Chloro-2,2,2-trifluoroacetophenone 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD10823-1GEA 23.9 EUR
BLDPBD10823-1G BLDPBD10823-5G BLDPBD10823-10G BLDPBD10823-250MG
4'-Chloro-2,2,2-trifluoroacetophenone 97%
4'-Chloro-2,2,2-trifluoroacetophenone

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₄ClF₃O
MW: 208,5700 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00018686
CAS-numero: 321-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR