6-Bromo-4-fluoro-1H-indazole 98%

BLDPBD108486-25GEA 1230 EUR
BLDPBD108486-25G BLDPBD108486-5G BLDPBD108486-1G BLDPBD108486-100MG BLDPBD108486-250MG
6-Bromo-4-fluoro-1H-indazole 98%
6-Bromo-4-fluoro-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₄BrFN₂
MW: 215,0200 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07781444
CAS-numero: 885520-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR