7H-Benzo[c]carbazole 97%

BLDPBD110615-1GEA 26 EUR
BLDPBD110615-1G BLDPBD110615-250MG BLDPBD110615-100MG BLDPBD110615-10G BLDPBD110615-5G BLDPBD110615-25G
7H-Benzo[c]carbazole 97%
7H-Benzo[c]carbazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₆H₁₁N
MW: 217,27 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00215940
CAS-numero: 205-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR