7-Hydroxynaphthalene-1-carbonitrile 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD110638-250MGEA 89.4 EUR
BLDPBD110638-250MG BLDPBD110638-100MG BLDPBD110638-1G
7-Hydroxynaphthalene-1-carbonitrile 97%
7-Hydroxynaphthalene-1-carbonitrile
Kaava: C₁₁H₇NO
MW: 169,179 g/mol
MDL numero: MFCD03695467
CAS-numero: 19307-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR