7-Methyl-1-tetralone 98%

BLDPBD110826-5GEA 73.8 EUR
BLDPBD110826-5G BLDPBD110826-1G BLDPBD110826-250MG BLDPBD110826-10G BLDPBD110826-100G BLDPBD110826-25G
7-Methyl-1-tetralone 98%
7-Methyl-1-tetralone

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₁H₁₂O
MW: 160,22 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00087600
CAS-numero: 22009-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR