4-Methylsalicylaldehyde 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD11145-25GEA 144 EUR
BLDPBD11145-25G BLDPBD11145-5G BLDPBD11145-250MG BLDPBD11145-100G BLDPBD11145-10G BLDPBD11145-1G
4-Methylsalicylaldehyde 98%
4-Methylsalicylaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₈H₈O₂
MW: 136,15 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00799550
CAS-numero: 698-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR