Methyl-4-amino-3-methoxybenzoate 98%

BLDPBD11301-25GEA 96.7 EUR
BLDPBD11301-25G BLDPBD11301-1G BLDPBD11301-10G BLDPBD11301-100G BLDPBD11301-5G
Methyl-4-amino-3-methoxybenzoate 98%
Methyl-4-amino-3-methoxybenzoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₉H₁₁NO₃
MW: 181,1900 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00017203
CAS-numero: 41608-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR