3'-Fluoro-5'-(trifluoromethyl)acetophenone 95%

BLDPBD11512-250MGEA 8.4 EUR
BLDPBD11512-250MG BLDPBD11512-0.025G BLDPBD11512-25G BLDPBD11512-1G BLDPBD11512-5G
3'-Fluoro-5'-(trifluoromethyl)acetophenone 95%
3'-Fluoro-5'-(trifluoromethyl)acetophenone

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₉H₆F₄O
MW: 206,1370 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00061250
CAS-numero: 202664-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR