4-Amino-N-methylbenzamide 98%

BLDPBD115401-10GEA 9.4 EUR
BLDPBD115401-10G BLDPBD115401-100G BLDPBD115401-25G
4-Amino-N-methylbenzamide 98%
4-Amino-N-methylbenzamide
Kaava: C₈H₁₀N₂O
MW: 150,1778 g/mol
MDL numero: MFCD00665792
CAS-numero: 6274-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR