Benzyl butyrate 98%

BLDPBD116122-100GEA 9.4 EUR
BLDPBD116122-100G BLDPBD116122-1000G BLDPBD116122-500G
Benzyl butyrate 98%
Benzyl butyrate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₁H₁₄O₂
MW: 178,23 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00027133
CAS-numero: 103-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR