Bis(triethoxysilyl)methane 97%

BLDPBD117251-5GEA 33.3 EUR
BLDPBD117251-5G BLDPBD117251-25G
Bis(triethoxysilyl)methane 97%
Bis(triethoxysilyl)methane

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₃H₃₂O₆Si₂
MW: 340,56 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00270971
CAS-numero: 18418-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR