3-Allylsalicylaldehyde 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD11823-5GEA 111 EUR
BLDPBD11823-5G BLDPBD11823-1G BLDPBD11823-250MG
3-Allylsalicylaldehyde 95%
3-Allylsalicylaldehyde

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₀H₁₀O₂
MW: 162,19 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00037364
CAS-numero: 24019-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR