3-hydroksi-4-metoksikanelihappo 98%

BLDPBD11852-1GEA 13.6 EUR
BLDPBD11852-0.025G BLDPBD11852-1G BLDPBD11852-5G BLDPBD11852-25G
3-hydroksi-4-metoksikanelihappo 98%
3-hydroksi-4-metoksikanelihappo

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₀H₁₀O₄
MW: 194,184 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004391
CAS-numero: 537-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR