Bosentan monohydrate 98%

BLDPBD118644-1GEA 59.3 EUR
BLDPBD118644-1G BLDPBD118644-250MG BLDPBD118644-5G BLDPBD118644-100MG
Bosentan monohydrate 98%
Bosentan monohydrate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₂₇H₂₉N₅O₆S·H₂O
MW: 551,62 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09751188
CAS-numero: 157212-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR