9,10-Dihydroacridine 98%

BLDPBD119964-25GEA 518 EUR
BLDPBD119964-25G BLDPBD119964-1G BLDPBD119964-0.025 BLDPBD119964-5G BLDPBD119964-250MG
9,10-Dihydroacridine 98%
9,10-Dihydroacridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₃H₁₁N
MW: 181,24 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00022256
CAS-numero: 92-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR