Thioacetazone 98%

BLDPBD12005-250MGEA 30.2 EUR
BLDPBD12005-250MG BLDPBD12005-1G BLDPBD12005-5G
Thioacetazone 98%
Thioacetazone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₀H₁₂N₄OS
MW: 236,2935 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00022157
CAS-numero: 104-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR