Choline iodide 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD121109-10GEA 23.9 EUR
BLDPBD121109-10G BLDPBD121109-25G BLDPBD121109-100G BLDPBD121109-5G
Choline iodide 98%
Choline iodide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₅H₁₄INO
MW: 231,08 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011900
CAS-numero: 17773-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR