1-(4-Nitrophenyl)piperazine 98%

BLDPBD12122-5GEA 10.4 EUR
BLDPBD12122-5G BLDPBD12122-100G BLDPBD12122-25G
1-(4-Nitrophenyl)piperazine 98%
1-(4-Nitrophenyl)piperazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₀H₁₃N₃O₂
MW: 207,23 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005961
CAS-numero: 6269-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR