5-Pyrimidineboronic acid pinacol ester 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD12251-1GEA 16.7 EUR
BLDPBD12251-1G BLDPBD12251-10G BLDPBD12251-5G BLDPBD12251-100G BLDPBD12251-250MG BLDPBD12251-25G
5-Pyrimidineboronic acid pinacol ester 97%
5-Pyrimidineboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₀H₁₅BN₂O₂
MW: 206,05 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD05155223
CAS-numero: 321724-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR