Deoxyanisoin 95%
|
|
BLDPBD123839-25GEA
74.9
EUR
BLDPBD123839-25G
BLDPBD123839-100G
BLDPBD123839-1G
BLDPBD123839-5G
Deoxyanisoin 95%
Deoxyanisoin
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₆H₁₆O₃ MW: 256,30 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00008406 CAS-numero: 120-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...