Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate 97%
|
|
BLDPBD124573-5GEA
135
EUR
BLDPBD124573-5G
BLDPBD124573-1G
BLDPBD124573-10G
BLDPBD124573-250MG
BLDPBD124573-100MG
Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate 97%
Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₃H₁₇NO₄ MW: 251,28 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00023506 CAS-numero: 1149-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...