Methoxy(dimethyl)-n-octylsilane 96% (by GC)

BLDPBD125597-1GEA 17.7 EUR
BLDPBD125597-1G BLDPBD125597-100G BLDPBD125597-25G BLDPBD125597-5G
Methoxy(dimethyl)-n-octylsilane 96% (by GC)
Methoxy(dimethyl)-n-octylsilane

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₁H₂₆OSi
MW: 202,41 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00043322
CAS-numero: 93804-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR