2-Amino-3-formylchromone 95%

BLDPBD12636-1GEA 87.3 EUR
BLDPBD12636-1G BLDPBD12636-5G BLDPBD12636-250MG
2-Amino-3-formylchromone 95%
2-Amino-3-formylchromone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₀H₇NO₃
MW: 189,1675 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00191735
CAS-numero: 61424-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR