7-Hydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one hydrate(2:3) 99%

BLDPBD127505-1GEA 7.3 EUR
BLDPBD127505-1G
7-Hydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one hydrate(2:3) 99%
7-Hydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one hydrate(2:3)
Kaava: C₃₀H₃₈O₂₁
MW: 734,61 g/mol
MDL numero: MFCD27981444
CAS-numero: 66778-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR