Dimethyl-4-nitrophthalate 97%
|
|
BLDPBD12909-1000GEA
444
EUR
BLDPBD12909-0.5G
BLDPBD12909-1000G
BLDPBD12909-25G
BLDPBD12909-100G
BLDPBD12909-500G
BLDPBD12909-10G
Dimethyl-4-nitrophthalate 97%
Dimethyl-4-nitrophthalate
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₀H₉NO₆ MW: 239,19 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00017191 CAS-numero: 610-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...