Ferroseeni 98%

BLDPBD130319-25GEA 7.3 EUR
BLDPBD130319-0.025 BLDPBD130319-25G
Ferroseeni 98%
Ferroseeni

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₀Fe
MW: 186,04 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001427
CAS-numero: 102-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR