N-Boc-4-bromoaniline 98%

BLDPBD13220-5GEA 10.4 EUR
BLDPBD13220-5G BLDPBD13220-1G BLDPBD13220-25G BLDPBD13220-10G BLDPBD13220-500G BLDPBD13220-100G
N-Boc-4-bromoaniline 98%
N-Boc-4-bromoaniline

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₁H₁₄BrNO₂
MW: 272,14 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01006612
CAS-numero: 131818-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR