Guanosine 5'-triphosphate disodium salt 89%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD132474-25GEA 629 EUR
BLDPBD132474-25G BLDPBD132474-5G
Guanosine 5'-triphosphate disodium salt 89%
Guanosine 5'-triphosphate disodium salt

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₁₀H₁₄N₅Na₂O₁₄P₃
MW: 567,147 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00083629
CAS-numero: 56001-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR