(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD132903-25GEA 148 EUR
BLDPBD132903-25G BLDPBD132903-5G BLDPBD132903-10G BLDPBD132903-1G
(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%
(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Kaava: C₁₀H₁₁NO₂
MW: 177,1998 g/mol
MDL numero: MFCD00191496
CAS-numero: 67123-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR