Heptanoyylikloridi 99%

BLDPBD133062-25GEA 10.4 EUR
BLDPBD133062-25G
Heptanoyylikloridi 99%
Heptanoyylikloridi

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₁₃ClO
MW: 148,63 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000763
CAS-numero: 2528-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR