1-Boc-4-cyanopiperidine 98%
|
|
BLDPBD13437-1GEA
6.3
EUR
BLDPBD13437-1G
BLDPBD13437-5G
BLDPBD13437-10G
BLDPBD13437-100G
BLDPBD13437-25G
BLDPBD13437-500G
1-Boc-4-cyanopiperidine 98%
1-Boc-4-cyanopiperidine
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₁₁H₁₈N₂O₂ MW: 210,27 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD01861223 CAS-numero: 91419-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...