Lithium dihydrogen orthophosphate 99%

BLDPBD136459-25GEA 7.3 EUR
BLDPBD136459-25G BLDPBD136459-1000G BLDPBD136459-100G BLDPBD136459-500G
Lithium dihydrogen orthophosphate 99%
Lithium dihydrogen orthophosphate

Storage condition: Inert atmosphere, Room Temperature

Kaava: H₂LiO₄P
MW: 103,93 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016188
CAS-numero: 13453-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR