Loxoprofen 97%

BLDPBD136713-10GEA 22.9 EUR
BLDPBD136713-10G BLDPBD136713-25G BLDPBD136713-1G BLDPBD136713-100G BLDPBD136713-5G
Loxoprofen 97%
Loxoprofen

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₅H₁₈O₃
MW: 246,31 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00864331
CAS-numero: 68767-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR