3-Biphenylboronic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD13795-5GEA 22.9 EUR
BLDPBD13795-5G BLDPBD13795-25G BLDPBD13795-10G BLDPBD13795-100G BLDPBD13795-1G
3-Biphenylboronic acid 98%
3-Biphenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₂H₁₁BO₂
MW: 198,03 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01318102
CAS-numero: 5122-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR